Läs mer
6-(5-Chloropyridin-2-yl)- 6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5,7-dione, Mikromol™
Produktkod. 30052661
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052661 25 mg
17940436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052661 Leverantör Mikromol Leverantörsnummer MM0149.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-(5-Chloropyridin-2-yl)- 6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5,7-dione
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-(5-Chloropyridin-2-yl)- 6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5,7-dione
CAS 43200-82-4
Analyt- eller komponentnamn 6-(5-Chloropyridin-2-yl)- 6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5,7-dione
Frakt skick Room Temperature
Molekylformel C11 H5 Cl N4 O2
Certifikat och efterlevnad ISO 9001
Synonym 6-(5-Chloropyridin-2-yl)- 6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5,7-dione
InChI formel InChI=1S/C11H5ClN4O2/c12-6-1-2-7(15-5-6)16-10(17)8-9(11(16)18)14-4-3-13-8/h1-5H
LEDER Clc1ccc(nc1)N2C(=O)c3nccnc3C2=O
IUPAC-namn 6-(5-chloropyridin-2-yl)pyrrolo[3,4-b]pyrazine-5,7-dione
Molekylvikt (g/mol) 260.64
Kvantitet 25 mg
Formel vikt 260.0101 g/mol