missing translation for 'onlineSavingsMsg'
Learn More

6(5H)-Phenanthridinone, 96%, Thermo Scientific Chemicals

Catalog Number p-7028064
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1015-89-0
C13H9NO
195.221
MFCD00004988
RZFVLEJOHSLEFR-UHFFFAOYSA-N
6 5h-phenanthridinone, phenanthridin-6 5h-one, 6-phenanthridinol, phenanthridone, 6-5h-phenanthridinone, 6-phenanthridone, 6-phenanthridinone, 6 5h-phenanthridone, phenanthridin-6-ol, phenantridone
1853
CHEBI:75292
5H-phenanthridin-6-one
C1=CC=C2C(=C1)C3=CC=CC=C3NC2=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1015-89-0
C13H9NO
195.221
MFCD00004988
RZFVLEJOHSLEFR-UHFFFAOYSA-N
6 5h-phenanthridinone, phenanthridin-6 5h-one, 6-phenanthridinol, phenanthridone, 6-5h-phenanthridinone, 6-phenanthridone, 6-phenanthridinone, 6 5h-phenanthridone, phenanthridin-6-ol, phenantridone
1853
CHEBI:75292
5H-phenanthridin-6-one
C1=CC=C2C(=C1)C3=CC=CC=C3NC2=O

6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

Solubility
Soluble in DMSO (5 mg/ml). Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 288°C to 294°C
Quantity 1 g
Beilstein 140641
Solubility Information Soluble in DMSO (5mg/ml). Insoluble in water.
Formula Weight 195.22
Percent Purity 96%
Chemical Name or Material 6(5H)-Phenanthridinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'