Läs mer
6,6’-Dibromoindigo, TRC
Produktkod. 30351791
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30351791 500 mg
1 options
Produktkod. 30351791 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD270675500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6,6'-Dibromoindigo
CAS 19201-53-7
Molekylformel C16 H8 Br2 N2 O2
Kvantitet 500 mg
Synonym (6CI) 6,6'-Dibromoindigotin, (7CI,8CI) 6,6'-Dibromo[δ2,2'-Biindoline]-3,3'-dione, 6-Bromo-2-(6-bromo-1,3-dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one, 6,6'-Dibromoindigotin, C.I. 75800, C.I. Natural Violet 1, Murex Purple, Purple of the Ancients, Royal Purple, Tyrian Purple
InChI formel InChI=1S/C16H8Br2N2O2/c17-7-1-3-9-11(5-7)19-13(15(9)21)14-16(22)10-4-2-8(18)6-12(10)20-14/h1-6,19,21H
LEDER Oc1c([nH]c2cc(Br)ccc12)C3=Nc4cc(Br)ccc4C3=O
IUPAC-namn 6-bromo-2-(6-bromo-3-hydroxy-1H-indol-2-yl)indol-3-one
Molekylvikt (g/mol) 420.055
Formel vikt 417.895