Läs mer
6,7,8,9-Tetrahydro Carvedilol, TRC
Produktkod. 30409743
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409743 25 mg
30424698 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30409743 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29335025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246820-73-4
Molekylformel C24 H30 N2 O4
Molekylvikt (g/mol) 410.51
Synonym 1-[2-(2-Methoxyphenoxy)ethylamino]-3-(6,7,8,9-tetrahydro-5H-carbazol-4-yloxy)propan-2-ol
IUPAC-namn 1-[2-(2-methoxyphenoxy)ethylamino]-3-(6,7,8,9-tetrahydro-5H-carbazol-4-yloxy)propan-2-ol
LEDER COc1ccccc1OCCNCC(O)COc2cccc3[nH]c4CCCCc4c23
Specifikationer
Kemiskt namn eller material 6,7,8,9-Tetrahydro Carvedilol
Kvantitet 25 mg
InChI formel InChI=1S/C24H30N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h4-6,9-12,17,25-27H,2-3,7-8,13-16H2,1H3
Formel vikt 410.2206