Läs mer
6,7-Bis(2-chloroethoxy)-N-(3-ethynylphenyl)-4-quinazolinamine Hydrochloride, TRC
Produktkod. 30329902
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30329902 25 mg
1 options
Produktkod. 30329902 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB41870025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6,7-Bis(2-chloroethoxy)-N-(3-ethynylphenyl)-4-quinazolinamin
CAS 183320-00-5
Molekylformel C20 H17 Cl2 N3 O2 . Cl H
Kvantitet 25 mg
Synonym 4-Quinazolinamine, 6,7-bis(2-chloroethoxy)-N-(3-ethynylphenyl)-, hydrochloride (1:1), 4-Quinazolinamine, 6,7-bis(2-chloroethoxy)-N-(3-ethynylphenyl)-, monohydrochloride (9CI), 6,7-Bis(2-chloroethoxy)-N-(3-ethynylphenyl)-4-quinazolinamine hydrochloride, 6,7-Bis(O-desmethoxyethyl)-6,7-bis(O-chloroethyl) erlotinib hydrochloride, 6,7-Bis(O-desmethoxyethyl)-6,7-bis(O-chloroethyl)erlotinib hydrochloride, 6,7-chloroethyl erlotinib hydrochloride, 6,7-Bis(2-chloroethoxy)-N-(3-ethynylphenyl) quinazolin-4-amine hydrochloride
InChI formel InChI=1S/C20H17Cl2N3O2.ClH/c1-2-14-4-3-5-15(10-14)25-20-16-11-18(26-8-6-21)19(27-9-7-22)12-17(16)23-13-24-20;/h1,3-5,10-13H,6-9H2,(H,23,24,25);1H
LEDER Cl.ClCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCCl
IUPAC-namn 6,7-bis(2-chloroethoxy)-N-(3-ethynylphenyl)quinazolin-4-amine;hydrochloride
Molekylvikt (g/mol) 438.73
Formel vikt 437.0465