Läs mer
6,7-Dehydro-17alpha-acetoxy Progesterone, TRC
Produktkod. 30012324
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012324 25 mg
30004243 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012324 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22909025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 425-51-4
Molekylformel C23H30O4
Molekylvikt (g/mol) 370.48
Synonym Pregna-4,6-diene-3,20-dione, 17-(acetyloxy)-, Pregna-4,6-diene-3,20-dione, 17-hydroxy-, acetate (6CI,8CI), 17-(Acetyloxy)pregna-4,6-diene-3,20-dione, 17-Acetoxypregna-4,6-diene-3,20-dione, 17α-Acetoxypregn-4,6-diene-3,20-dione, 17α-Acetoxypregna-4,6-diene-3,20-dione, 17α-Hydroxy-6-dehydroprogesterone acetate, 6-Dehydro-17α-acetoxyprogesterone, Melengestro acetate, NSC 77897, Pregna-4,6-dien-17-ol-3,20-dione acetate, δ6-Dehydro-17α-acetoxyprogesterone, 6,7-Didehydro-17α-acetoxyprogesterone
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 6,7-Dehydro-17a-acetoxy Progesterone
Kvantitet 25 mg
InChI formel InChI=1S/C23H30O4/c1-14(24)23(27-15(2)25)12-9-20-18-6-5-16-13-17(26)7-10-21(16,3)19(18)8-11-22(20,23)4/h5-6,13,18-20H,7-12H2,1-4H3/t18-,19+,20+,21+,22+,23+/m1/s1
Formel vikt 370.2144