Läs mer
6,7-Dehydro Estradiol, TRC
Produktkod. 30179746
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179746 100 mg
30180181 5 mg
30179894 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7291-41-0
Molekylformel C18 H22 O2
Molekylvikt (g/mol) 270.37
Synonym Estra-1,3,5(10),6-tetraene-3,17-beta-diol, 6-Dehydroestradiol, Estra-1,3,5(10)-6-tetraen-3,17-beta-diol, NSC 57430, delta-6,7-Estradiol, 6,7-Didehydroestradiol
IUPAC-namn (8R,9S,13S,14S,17S)-13-methyl-8,9,11,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol
LEDER C[C@]12CC[C@H]3[C@@H](C=Cc4cc(O)ccc34)[C@@H]1CC[C@@H]2O
Specifikationer
Kemiskt namn eller material 1,3,5(10),6-estratetraen-3,17beta-diol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,14-17,19-20H,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1
Formel vikt 270.162
Procent renhet >95%