Läs mer
6,7-Dehydro Ethynyl Estradiol, TRC
Produktkod. 30132969
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132969 25 mg
17974195 10 mg
17972235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132969 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22960525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67703-68-8
Molekylformel C20H22O2
Molekylvikt (g/mol) 294.39
Synonym 19-Nor-17alpha-pregna-1,3,5(10),6-tetraen-20-yne-3,17-diol, Ethinylestradiol Imp. I (EP), 6,7-Didehydroethinylestradiol
IUPAC-namn (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-9,11,12,14,15,16-hexahydro-8H-cyclopenta[a]phenanthrene-3,17-diol
LEDER C[C@]12CC[C@H]3[C@@H](C=Cc4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material 19-Nor-17-alpha-pregna-1,3,5(10),6-tetraen-20-yne-3,17-diol (6,7-Didehydroethinylestradiol)
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C20H22O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,4-7,12,16-18,21-22H,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1
Formel vikt 294.162
Procent renhet >95%