Läs mer
6(7)-Dehydro Norgestrel, TRC
Produktkod. 30010929
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010929 10 mg
30007372 5 mg
30003280 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010929 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22992010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51087-61-7
Molekylformel C21H26O2
Molekylvikt (g/mol) 310.43
Synonym 13-Ethyl-17-hydroxy-18,19-dinor-17alpha-pregna-4,6-dien-20-yn-3-one, Levonorgestrel Imp. M (Pharmeuropa), Delta6-Levonorgestrel, 6,7-Didehydrolevonorgestrel
IUPAC-namn (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,8,9,10,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
LEDER CC[C@]12CC[C@H]3[C@@H](C=CC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material 13-Ethyl-17-hydroxy-18,19-dinor-17alpha-pregna-4,6-dien-20-yn-3-one (delta 6-Levonorgestrel; 6,7-Didehydrolevonorgestrel)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C21H26O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,5,7,13,16-19,23H,3,6,8-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
Formel vikt 310.1933
Procent renhet >95%