Läs mer
6,7-Dichloro-1-tetralone, TRC
Produktkod. 30376495
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376495 500 mg
30368962 50 mg
30377113 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30376495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD439878500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 25095-57-2
Molekylformel C10H8OCl2
Molekylvikt (g/mol) 215.08
LEDER Clc1cc2CCCC(=O)c2cc1Cl
Specifikationer
Kemiskt namn eller material 6,7-Dichloro-1-tetralone
Kvantitet 500 mg
InChI formel InChI=1S/C10H8Cl2O/c11-8-4-6-2-1-3-10(13)7(6)5-9(8)12/h4-5H,1-3H2
Formel vikt 213.9952