Läs mer
6,7-Dihydroxyflavone, TRC
Produktkod. 30180054
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180054 1 mg
30179858 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180054 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6790081MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38183-04-9
Molekylformel C15 H10 O4
Molekylvikt (g/mol) 254.24
IUPAC-namn 6,7-dihydroxy-2-phenyl-chromen-4-one
LEDER Oc1cc2OC(=CC(=O)c2cc1O)c3ccccc3
Specifikationer
Kemiskt namn eller material 6,7-Dihydroxyflavone
Kvantitet 1 mg
InChI formel InChI=1S/C15H10O4/c16-11-7-14(9-4-2-1-3-5-9)19-15-8-13(18)12(17)6-10(11)15/h1-8,17-18H
Formel vikt 254.0579