missing translation for 'onlineSavingsMsg'
Läs mer

6-Acetamidohexanoic Acid, TRC

Produktkod. 30328705
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30328705 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30328705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA158410100G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 6-Acetamidohexanoic Acid
CAS 57-08-9
Molekylformel C8H15NO3
Kvantitet 100 g
Synonym 6-(Acetylamino)hexanoic Acid, 6-Acetamidocaproic Acid, Acemin, Acetaminocaproic Acid, Acexamic Acid, CY 153, N-Acetyl-6-aminohexanoic Acid, AACA, N-Acetyl-epsilon-aminocaproic alphacid, NSC 12945, epsilon-Acetamidocaproic Acid
InChI formel InChI=1S/C8H15NO3/c1-7(10)9-6-4-2-3-5-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)
LEDER CC(=O)NCCCCCC(=O)O
IUPAC-namn 6-acetamidohexanoic acid
Molekylvikt (g/mol) 173.21
Formel vikt 173.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.