Läs mer
6-Allyl-8Beta-carboxyergoline Methyl Ester, TRC
Produktkod. 30317585
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30317585 5 mg
1 options
Produktkod. 30317585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5496155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Allyl-8beta-carboxyergoline Methyl Ester
CAS 72821-79-5
Molekylformel C19H22N2O2
Kvantitet 5 mg
Synonym (8beta)-6-(2-Propen-1-yl)-ergoline-8-carboxylic Acid Methyl Ester, 6-(2-Propenyl)dihydrolysergic Acid Methyl Ester, Indolo[4,3-fg]quinoline, ergoline-8-carboxylic Acid Deriv.
InChI formel InChI=1S/C19H22N2O2/c1-3-7-21-11-13(19(22)23-2)8-15-14-5-4-6-16-18(14)12(10-20-16)9-17(15)21/h3-6,10,13,15,17,20H,1,7-9,11H2,2H3/t13-,15-,17-/m1/s1
LEDER COC(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(CC=C)C1
IUPAC-namn methyl (6aR,9R,10aR)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxylate
Molekylvikt (g/mol) 310.39
Formel vikt 310.1681