Läs mer
6-Amidino-2-naphthol Methanesulfonate, TRC
Produktkod. 30310691
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30310691 100 mg
1 options
Produktkod. 30310691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA510060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Amidino-2-naphthol Methanesulfonate
CAS 82957-06-0
Molekylformel C11H10N2O . CH4SO3
Kvantitet 100 mg
Synonym 6-Amidino-2-Naphthol Methanesulfonic Acid
LEDER CS(=O)(=O)O.NC(=N)c1ccc2cc(O)ccc2c1
Formel vikt 282.0674