missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-hexanol, 94%, Thermo Scientific Chemicals

Catalog Number 10713914
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4048-33-3
C6H15NO
117.19
MFCD00008241
SUTWPJHCRAITLU-UHFFFAOYSA-N
6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
19960
6-aminohexan-1-ol
C(CCCO)CCN
This item is not returnable. View return policy
4048-33-3
C6H15NO
117.19
MFCD00008241
SUTWPJHCRAITLU-UHFFFAOYSA-N
6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
19960
6-aminohexan-1-ol
C(CCCO)CCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 92.0
CAS Max % 100.0
Melting Point 53°C to 59°C
Color Yellow to Brown
Boiling Point 233.0°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)6OH
Quantity 50 g
Beilstein 04, II, 748
Solubility Information Solubility in water: soluble
Formula Weight 117.19
Percent Purity 94%
Physical Form Crystals
Chemical Name or Material 6-Amino-1-hexanol
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-1-hexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'