Läs mer
6-Amino-1-methyl-5-nitrosouracil, TRC
Produktkod. 30321993
Change view
Kvantitet:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30321993 100 g
30316872 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30321993 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA616980100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Amino-1-methyl-5-nitrosouracil
CAS 6972-78-7
Molekylformel C5H6N4O3
Kvantitet 100 g
Synonym 6-Amino-1-methyl-5-nitroso-2,4-pyrimidinedione, 6-amino-1-methyl-5-nitroso-uracil, 6-Amino-1-methyl-5-nitrosopyrimidine-2,4-dione, NSC 62582
InChI formel InChI=1S/C5H6N4O3/c1-9-3(6)2(8-12)4(10)7-5(9)11/h6H2,1H3,(H,7,10,11)
LEDER CN1C(=C(N=O)C(=O)NC1=O)N
IUPAC-namn 6-amino-1-methyl-5-nitrosopyrimidine-2,4-dione
Molekylvikt (g/mol) 170.13
Formel vikt 170.044