missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-pyridinemethanol, 97%, Thermo Scientific™

Catalog Number p-7077337
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79651-64-2
C6H8N2O
124.14
YGMPMXTWKROXPP-UHFFFAOYSA-N
6-aminopyridin-2-yl methanol, 6-amino-2-pyridinemethanol, 2-aminopyridine-6-methanol, 6-amino-pyridin-2-yl-methanol, 2-pyridinemethanol, 6-amino, 2-amino-6-hydroxymethylpyridine, 6-aminopyridine-2-methanol, 6-amino-2-pyridinyl methanol, 2-amino-6-hydroxymethyl pyridine, 2-hydroxymethyl-6-aminopyridine
11182606
(6-aminopyridin-2-yl)methanol
C1=CC(=NC(=C1)N)CO
This item is not returnable. View return policy
79651-64-2
C6H8N2O
124.14
YGMPMXTWKROXPP-UHFFFAOYSA-N
6-aminopyridin-2-yl methanol, 6-amino-2-pyridinemethanol, 2-aminopyridine-6-methanol, 6-amino-pyridin-2-yl-methanol, 2-pyridinemethanol, 6-amino, 2-amino-6-hydroxymethylpyridine, 6-aminopyridine-2-methanol, 6-amino-2-pyridinyl methanol, 2-amino-6-hydroxymethyl pyridine, 2-hydroxymethyl-6-aminopyridine
11182606
(6-aminopyridin-2-yl)methanol
C1=CC(=NC(=C1)N)CO
Melting Point 138.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 6-Amino-2-pyridinemethanol
compliance-icons
Product Identifier
  • 6-Amino-2-pyridinemethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'