Learn More
6-Aminonaphthalen-1-ol, 98%, BLD Pharm™
Product Code. 30944925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30944925 5 g 5g
30943680 1 g 1g
30930710 250 mg 250mg
30936265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30944925 Supplier BLD Pharm Supplier No. BD128585g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 23894-12-4
Molecular Formula C10H9NO
MDL Number MFCD00059543
Quantity 5 g
SMILES OC1=C2C=CC(N)=CC2=CC=C1
Molecular Weight (g/mol) 159.18
Percent Purity 98%
Chemical Name or Material 6-Aminonaphthalen-1-ol