Läs mer
6-Azabicyclo[3.2.1]octane Hydrochloride, TRC
Produktkod. 30424343
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424343 250 mg
30425996 500 mg
30426375 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424343 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS684588250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40314-13-4
Molekylformel C7H13N . HCl
Molekylvikt (g/mol) 147.64
Synonym 6-Azabicyclo[3.2.1]octane, hydrochloride (1:1) (ACI), 6-Azabicyclo[3.2.1]octane, hydrochloride (7CI, 9CI), 6-Azabicyclo[3.2.1]octane hydrochloride
IUPAC-namn 6-azabicyclo[3.2.1]octane;hydrochloride
LEDER Cl.C1CC2CNC(C1)C2
Specifikationer
Kemiskt namn eller material 6-Azabicyclo[3.2.1]octane Hydrochloride
Kvantitet 250 mg
InChI formel InChI=1S/C7H13N.ClH/c1-2-6-4-7(3-1)8-5-6;/h6-8H,1-5H2;1H
Formel vikt 147.0815