Läs mer
Tocris Bioscience™ 6-Bnz-cAMP sodium salt
Cell permeable cAMP analog
Brand: Tocris Bioscience™ 5255/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
6-Bnz-cAMP sodium salt is a cell permeable cAMP analog; selectively activates cAMP-dependent PKA but not Epac signaling pathways. Acts synergistically with 8-CPT-2Me-cAMP (Cat. No. 1645) to inhibit vascular smooth muscle cell proliferation.Specifikationer
| 98% | |
| 1135306-29-4 | |
| 1 mg |
| N-Benzoyladenosine cyclic 3 ',5 '-(hydrogen phosphate) monosodium salt | |
| -20°C |
Produkttitel
For Research Use Only