missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-(Boc-amino)-5-bromoquinoline, 95%

Brand:  Alfa Aesar™ H63551.06

Additional Details : CAS Number : 889650-18-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450299

  • 12967.81 SEK / 5g


Estimated Shipment date
from Supplier 16-11-2018
Add to basket

Specifications

Chemical Identifiers

889650-18-4
323.19
6-boc-amino-5-bromoquinoline, tert-butyl 5-bromoquinolin-6-yl carbamate, tert-butyl n-5-bromoquinolin-6-yl carbamate
tert-butyl N-(5-bromoquinolin-6-yl)carbamate
C14H15BrN2O2
NDSMGXYVEQETDA-UHFFFAOYSA-N
86616929
CC(C)(C)OC(=O)NC1=C(C2=C(C=C1)N=CC=C2)Br

Specifications

6-(Boc-amino)-5-bromoquinoline
889650-18-4
14556821
NDSMGXYVEQETDA-UHFFFAOYSA-N
tert-butyl N-(5-bromoquinolin-6-yl)carbamate
86616929
Solid
5g
C14H15BrN2O2
6-boc-amino-5-bromoquinoline, tert-butyl 5-bromoquinolin-6-yl carbamate, tert-butyl n-5-bromoquinolin-6-yl carbamate
CC(C)(C)OC(=O)NC1=C(C2=C(C=C1)N=CC=C2)Br
323.19
323.18
95%
Videos
SDS