missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Boc-amino)hexanoic acid, 95%, Thermo Scientific Chemicals
752.66 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.
Solubility
Soluble in chloroform and ethyl acetate. Slightly soluble in water.
Notes
Store at -20°C. Keep container tightly closed. With good ventilation. Protect form heat and store away from oxidizing agent.
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.
Solubility
Soluble in chloroform and ethyl acetate. Slightly soluble in water.
Notes
Store at -20°C. Keep container tightly closed. With good ventilation. Protect form heat and store away from oxidizing agent.
Specifications
Melting Point | 35°C to 40°C |
Color | White |
Linear Formula | (CH3)3COCONH(CH2)5CO2H |
Quantity | 5 g |
Beilstein | 2049561 |
Solubility Information | Soluble in chloroform and ethyl acetate. Slightly soluble in water. |
Formula Weight | 231.29 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 6-(Boc-amino)hexanoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'