missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2(1H)-quinolinone, 96%, Thermo Scientific Chemicals

Catalog Number p-7035874
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1810-66-8
C9H6BrNO
224.06
MFCD08703169
YLAFBGATSQRSTB-UHFFFAOYSA-N
6-bromoquinolin-2 1h-one, 6-bromo-2 1h-quinolone, 6-bromo-2 1h-quinolinone, 6-bromoquinolin-2-ol, 6-bromo-2-quinolone, 6-bromo-2-hydroxyquinoline, 6-bromo-1,2-dihydroquinolin-2-one, 2 1h-quinolinone, 6-bromo, 2-hydroxy-6-bromoquinoline, 6-bromocarbostyril
12378943
6-bromo-1H-quinolin-2-one
BrC1=CC=C2NC(=O)C=CC2=C1
This item is not returnable. View return policy
1810-66-8
C9H6BrNO
224.06
MFCD08703169
YLAFBGATSQRSTB-UHFFFAOYSA-N
6-bromoquinolin-2 1h-one, 6-bromo-2 1h-quinolone, 6-bromo-2 1h-quinolinone, 6-bromoquinolin-2-ol, 6-bromo-2-quinolone, 6-bromo-2-hydroxyquinoline, 6-bromo-1,2-dihydroquinolin-2-one, 2 1h-quinolinone, 6-bromo, 2-hydroxy-6-bromoquinoline, 6-bromocarbostyril
12378943
6-bromo-1H-quinolin-2-one
BrC1=CC=C2NC(=O)C=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >200°C
Quantity 1 g
Formula Weight 224.05
Percent Purity 96%
Chemical Name or Material 6-Bromo-2(1H)-quinolinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'