missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-benzothiazolinone, 98%, ACROS Organics™

574.00 SEK - 1705.40 SEK

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.08
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
IUPAC Name 6-bromo-3H-1,3-benzothiazol-2-one
SMILES C1=CC2=C(C=C1Br)SC(=O)N2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021971
SDS View Documents
Acros Organics
339880010
1g Glass bottle
574.00 SEK
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 2
Add to basket
 
10525691
SDS View Documents
Acros Organics
339880050
5g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

62266-82-4
230.08
HECJMTPEVWQFCY-UHFFFAOYSA-N
188444
C1=CC2=C(C=C1Br)SC(=O)N2
C7H4BrNOS
MFCD00239363
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
6-bromo-3H-1,3-benzothiazol-2-one

Specifications

6-Bromo-2-benzothiazolinone
Glass bottle
228°C to 232°C
98%
97.5
98%
MFCD00239363
Solubility in water: insoluble in water.,soluble in ethanol
C1=CC2=C(C=C1Br)SC(=O)N2
230.08
230.08
98%
Authentic
White
1g
62266-82-4
100.0
C7H4BrNOS
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
HECJMTPEVWQFCY-UHFFFAOYSA-N
6-bromo-3H-1,3-benzothiazol-2-one
188444
Crystalline Powder
SDS