missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chloro-1,8-naphthyridine, 96%, Thermo Scientific Chemicals

Catalog Number p-7033038
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
902837-40-5
C8H4BrClN2
243.488
MFCD07781152
KXSBJEOTLVRREX-UHFFFAOYSA-N
6-bromo-2-chloro-1,8 naphthyridine, 1,8-naphthyridine, 6-bromo-2-chloro, pubchem18342, ksc495s9r, abbypharma ap-14-10902, 6-bromo-2-chloropyridino 2,3-b pyridine, 6-bromanyl-2-chloranyl-1,8-naphthyridine
25631443
6-bromo-2-chloro-1,8-naphthyridine
C1=CC(=NC2=NC=C(C=C21)Br)Cl
This item is not returnable. View return policy
902837-40-5
C8H4BrClN2
243.488
MFCD07781152
KXSBJEOTLVRREX-UHFFFAOYSA-N
6-bromo-2-chloro-1,8 naphthyridine, 1,8-naphthyridine, 6-bromo-2-chloro, pubchem18342, ksc495s9r, abbypharma ap-14-10902, 6-bromo-2-chloropyridino 2,3-b pyridine, 6-bromanyl-2-chloranyl-1,8-naphthyridine
25631443
6-bromo-2-chloro-1,8-naphthyridine
C1=CC(=NC2=NC=C(C=C21)Br)Cl

6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 243.49
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-chloro-1,8-naphthyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'