missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 137°C to 138°C |
Quantity | 1 g |
Sensitivity | Light sensitive |
Synonym | 6-bromo-2-methylpyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 6-bromo-2-methyl, pyrazolo 1,5-a pyrimidine,6-bromo-2-methyl |
Formula Weight | 212.05 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'