missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 95%

Brand:  Alfa Aesar™ H63284.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488216

  • 10374.25 SEK / 5g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

6-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
MFCD09954823

Specifications

6-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
5g
95%
277.16
Solid
MFCD09954823
Videos
SDS