missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole, 97%

Brand:  Alfa Aesar™ H63660.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15498426

  • 6488.72 SEK / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
334.21
MFCD28098698
6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole

Specifications

6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
97%
334.21
6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
Solid
5g
MFCD28098698
Videos
SDS