missing translation for 'onlineSavingsMsg'
Läs mer

6-Bromo-4-hydroxyquinoline-3-carboxylic acid, 96%, Thermo Scientific™

Produktkod. 15400748
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15400748 5 g 5g
15401196 250 mg 250mg
15490738 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15400748 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H54441.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 98948-95-9
Molekylformel C10H6BrNO3
Molekylvikt (g/mol) 268.066
MDL-nummer MFCD01569278
InChI-nyckel GIUZUAUCCUFVKW-UHFFFAOYSA-N
Synonym 6-bromo-4-hydroxyquinoline-3-carboxylic acid, 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 6-bromo-4-hydroxy-3-quinolinecarboxylic acid, 4-hydroxy-6-bromoquinoline-3-carboxylic acid, 3-quinolinecarboxylic acid, 6-bromo-4-hydroxy, 3-quinolinecarboxylic acid, 6-bromo-1,4-dihydro-4-oxo, 6-bromo-4-hydroxyquinoline-3-carboxylicacid, 6-bromo-4-hydroxy-quinoline-3-carboxylic acid, 6-bromo-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, pubchem17804
PubChem CID 693203
IUPAC-namn 6-bromo-4-oxo-1H-quinoline-3-carboxylic acid
LEDER C1=CC2=C(C=C1Br)C(=O)C(=CN2)C(=O)O

Specifications

Kemiskt namn eller material 6-Bromo-4-hydroxyquinoline-3-carboxylic acid
Kvantitet 5 g
Löslighetsinformation Slightly soluble in water.
Formel vikt 268.06
Procent renhet 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.