Learn More
Alfa Aesar™ 6-Bromo-5-nitroisoquinoline, 95%
Brand:  Alfa Aesar™ H63861.MD
Additional Details : CAS Number : 850197-72-7 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15450799
  • 1291.96 SEK / 250mg
Estimated Shipment date
from Supplier 22-10-2018
Add to basket
Specifications
Chemical Identifiers
850197-72-7
253.055
COTVYJVDXKFNRB-UHFFFAOYSA-N
6-bromo-5-nitroisoquinoline
C9H5BrN2O2
MFCD08282782
34176644
C1=CC(=C(C2=C1C=NC=C2)[N+](=O)[O-])Br
Specifications
6-Bromo-5-nitroisoquinoline
95%
C9H5BrN2O2
COTVYJVDXKFNRB-UHFFFAOYSA-N
6-bromo-5-nitroisoquinoline
34176644
Solid
850197-72-7
250mg
MFCD08282782
C1=CC(=C(C2=C1C=NC=C2)[N+](=O)[O-])Br
253.055
253.05