Läs mer
6-Bromobenzene-1,2,4-triol, TRC
Produktkod. 30334298
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334298 25 mg
30335441 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30334298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB79583525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Bromobenzene-1,2,4-triol
CAS 1480325-12-9
Molekylformel C6H5BrO3
Kvantitet 25 mg
Synonym 6-Bromo-1,2,4-benzenetriol
InChI formel InChI=1S/C6H5BrO3/c7-4-1-3(8)2-5(9)6(4)10/h1-2,8-10H
LEDER Oc1cc(O)c(O)c(Br)c1
IUPAC-namn 6-bromobenzene-1,2,4-triol
Molekylvikt (g/mol) 205.01
Formel vikt 203.9422