missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole, 96%, Thermo Scientific Chemicals

Catalog Number p-3777663
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
52415-29-9
C8H6BrN
196.05
MFCD00238550
MAWGHOPSCKCTPA-UHFFFAOYSA-N
6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
676493
6-bromo-1H-indole
C1=CC(=CC2=C1C=CN2)Br
This item is not returnable. View return policy
52415-29-9
C8H6BrN
196.05
MFCD00238550
MAWGHOPSCKCTPA-UHFFFAOYSA-N
6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
676493
6-bromo-1H-indole
C1=CC(=CC2=C1C=CN2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 92.0°C to 96.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Formula Weight 196.05
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Bromoindole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'