missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromooxindole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 212.05 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromooxindole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'