Läs mer
6-Carboxyhexylphosphocholine p-Nitrophenyl Ester, TRC
Produktkod. 30367423
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30367423 25 mg
1 options
Produktkod. 30367423 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17846025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Carboxyhexylphosphocholine p-Nitrophenyl Ester
CAS 73785-43-0
Molekylformel C17 H27 N2 O8 P
Kvantitet 25 mg
Synonym Ethanaminium, 2-[[hydroxy[[6-(4-nitrophenoxy)-6-oxohexyl]oxy]phosphinyl]oxy]-N,N,N-trimethyl-, inner salt, 6-Carboxyhexylphosphocholine p-Nitrophenyl Ester
InChI formel InChI=1S/C17H27N2O8P/c1-19(2,3)12-14-26-28(23,24)25-13-6-4-5-7-17(20)27-16-10-8-15(9-11-16)18(21)22/h8-11H,4-7,12-14H2,1-3H3
LEDER C[N+](C)(C)CCOP(=O)([O-])OCCCCCC(=O)Oc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn [6-(4-nitrophenoxy)-6-oxohexyl] 2-(trimethylazaniumyl)ethyl phosphate
Molekylvikt (g/mol) 418.38
Formel vikt 418.1505