Läs mer
6-Chloro-1,2-benzisothiazol-3(2H)-one, TRC
Produktkod. 30365047
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365047 50 mg
30360392 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365047 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36462550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70-10-0
Molekylformel C7 H4 Cl N O S
Molekylvikt (g/mol) 185.63
Synonym 1,2-Benzisothiazol-3(2H)-one, 6-chloro-, 1,2-Benzisothiazolin-3-one, 6-chloro- (7CI,8CI), 6-Chloro-1,2-benzisothiazol-3(2H)-one, 6-Chloro-1,2-benzisothiazole-3-one, 6-Chloro-1,2-benzisothiazolin-3-one, 6-Chloro-1,2-benzoisothiazol-3-one, FER 1443, Landromil, Ticlaton, Ticlatone
IUPAC-namn 6-chloro-1,2-benzothiazol-3-one
LEDER Clc1ccc2C(=O)NSc2c1
Specifikationer
Kemiskt namn eller material 6-Chloro-1,2-benzisothiazol-3(2H)-one
Kvantitet 50 mg
InChI formel InChI=1S/C7H4ClNOS/c8-4-1-2-5-6(3-4)11-9-7(5)10/h1-3H,(H,9,10)
Formel vikt 184.9702