Läs mer
6-Chloro-1-benzofuran-2-carboxylic Acid, TRC
Produktkod. 30368440
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368440 250 mg
30363101 1 g
30357205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368440 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367265250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 442125-04-4
Molekylformel C9H5ClO3
Molekylvikt (g/mol) 196.59
Synonym 6-Chloro-2-benzofurancarboxylic Acid
IUPAC-namn 6-chloro-1-benzofuran-2-carboxylic acid
LEDER OC(=O)c1oc2cc(Cl)ccc2c1
Specifikationer
Kemiskt namn eller material 6-Chloro-1-benzofuran-2-carboxylic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C9H5ClO3/c10-6-2-1-5-3-8(9(11)12)13-7(5)4-6/h1-4H,(H,11,12)
Formel vikt 195.9927