missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-dihydroimidazo[1,2-b]pyridazine, 98%, Thermo Scientific™

Catalog Number 15438088
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
24439-00-7
C6H6ClN3
155.585
KSDBTFCPAJCBSO-UHFFFAOYSA-N
6-chloro-2,3-dihydroimidazo 1,2-b pyridazine, 6-chloro-2h,3h-imidazo 1,2-b pyridazine, 6-chloro-2,3-dihydro-imidazo 1,2-b pyridazine
83290716
6-chloro-2,3-dihydroimidazo[1,2-b]pyridazine
C1CN2C(=N1)C=CC(=N2)Cl
This item is not returnable. View return policy
24439-00-7
C6H6ClN3
155.585
KSDBTFCPAJCBSO-UHFFFAOYSA-N
6-chloro-2,3-dihydroimidazo 1,2-b pyridazine, 6-chloro-2h,3h-imidazo 1,2-b pyridazine, 6-chloro-2,3-dihydro-imidazo 1,2-b pyridazine
83290716
6-chloro-2,3-dihydroimidazo[1,2-b]pyridazine
C1CN2C(=N1)C=CC(=N2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 155.58
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6-Chloro-2,3-dihydroimidazo[1,2-b]pyridazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'