Läs mer
6-Chloro-2,3-dimethyl-1-phenyl-1H-benzo[d]imidazol-3-ium Chloride, TRC
Produktkod. 30135948
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135948 50 mg
30011402 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36723050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75524-10-6
Molekylformel C15 H14 Cl N2 . Cl
Molekylvikt (g/mol) 293.191
Synonym 5-chloro-1,2-dimethyl-3-phenyl-1H-Benzimidazolium Chloride, Clobazam impurity
IUPAC-namn 5-chloro-1,2-dimethyl-3-phenylbenzimidazol-3-ium;chloride
LEDER [Cl-].Cc1[n+](c2ccccc2)c3cc(Cl)ccc3n1C
Specifikationer
Kemiskt namn eller material 6-Chloro-2,3-dimethyl-1-phenyl-1H-benzo[d]imidazol-3-ium Chloride
Kvantitet 50 mg
InChI formel InChI=1S/C15H14ClN2.ClH/c1-11-17(2)14-9-8-12(16)10-15(14)18(11)13-6-4-3-5-7-13;/h3-10H,1-2H3;1H/q+1;/p-1
Formel vikt 292.053