missing translation for 'onlineSavingsMsg'
Läs mer

6-Chloro-2-fluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Produktkod. 11471420
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
10 g
Förpackningsstorlek:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11471420 10 g 10g
11461420 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11471420 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26444.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air and reducing agents. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 112641-64-2
Molekylformel C8H6ClFO2
Molekylvikt (g/mol) 188.582
MDL-nummer MFCD04115946
InChI-nyckel OADUJQWBRPRPQX-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-5-methoxybenzaldehyde, 6-chloro-2-fluoro-3-methoxy-benzaldehyde, 6-chloro-2-fluoro-3-methoxybenzaldehyde
PubChem CID 3580261
IUPAC-namn 6-chloro-2-fluoro-3-methoxybenzaldehyde
LEDER COC1=C(C(=C(C=C1)Cl)C=O)F

Specifikationer

Kemiskt namn eller material 6-Chloro-2-fluoro-3-methoxybenzaldehyde
Smältpunkt 95°C to 100°C
Kvantitet 10 g
Känslighet Air Sensitive
Formel vikt 188.59
Procent renhet 97%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.