Läs mer
6-Chloro-3-fluoro-2-iodopyridine, TRC
Produktkod. 30240834
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240834 10 mg
30240687 100 mg
30240665 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240834 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38297810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1211590-36-1
Molekylformel C5H2NFClI
Molekylvikt (g/mol) 257.43
IUPAC-namn 6-chloro-3-fluoro-2-iodopyridine
LEDER Fc1ccc(Cl)nc1I
Specifikationer
Kemiskt namn eller material 6-Chloro-3-fluoro-2-iodopyridine
Kvantitet 10 mg
InChI formel InChI=1S/C5H2ClFIN/c6-4-2-1-3(7)5(8)9-4/h1-2H
Formel vikt 256.8904