Läs mer
6-Chloro-3-hydroxyindolin-2-one, TRC
Produktkod. 30368387
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Kvantitet
30368387 100 mg
30367675 1 g
30357232 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30368387 Fornitore Toronto Research Chemicals N. del fornitore TRCC996370100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1251137-37-7
Molekylformel C8H6ClNO2
Molekylvikt (g/mol) 183.59
Synonym 6-Chloro-1,3-dihydro-3-hydroxy-2H-indol-2-one
IUPAC-namn 6-chloro-3-hydroxy-1,3-dihydroindol-2-one
LEDER OC1C(=O)Nc2cc(Cl)ccc12
Specifikationer
Kemiskt namn eller material 6-Chloro-3-hydroxyindolin-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C8H6ClNO2/c9-4-1-2-5-6(3-4)10-8(12)7(5)11/h1-3,7,11H,(H,10,12)
Formel vikt 183.0087