missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-iodo-4H-chromen-4-one, 97%, Thermo Scientific™

Catalog Number p-7031277
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
73220-39-0
C9H4ClIO2
306.483
MFCD09475515
GMONZFZWCHBAKU-UHFFFAOYSA-N
6-chloro-3-iodochromone, 6-chloro-3-iodo-4h-chromen-4-one, acmc-20alzd, 6-chloro-3-iodo-chromone, 6-chloro-3-iodo-4h-benzopyran-4-one, 3-iodo-6-chloro-4h-1-benzopyran-4-one
11666677
6-chloro-3-iodochromen-4-one
C1=CC2=C(C=C1Cl)C(=O)C(=CO2)I
This item is not returnable. View return policy
73220-39-0
C9H4ClIO2
306.483
MFCD09475515
GMONZFZWCHBAKU-UHFFFAOYSA-N
6-chloro-3-iodochromone, 6-chloro-3-iodo-4h-chromen-4-one, acmc-20alzd, 6-chloro-3-iodo-chromone, 6-chloro-3-iodo-4h-benzopyran-4-one, 3-iodo-6-chloro-4h-1-benzopyran-4-one
11666677
6-chloro-3-iodochromen-4-one
C1=CC2=C(C=C1Cl)C(=O)C(=CO2)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 143°C to 146°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 306.49
Percent Purity 97%
Chemical Name or Material 6-Chloro-3-iodo-4H-chromen-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'