missing translation for 'onlineSavingsMsg'
Läs mer

6-Kloro-3-(propan-2-yl)-1,2,3,4-tetrahydropyrimidin-2,4-dion, TRC

Produktkod. 30342272
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30342272 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342272 Supplier Toronto Research Chemicals Supplier No. TRCB428180500MG
 Begränsad artikel
Beräknad leverans: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 6-Chloro-3-(propan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
CAS 887581-47-7
Molekylformel C7H9ClN2O2
Kvantitet 500 mg
Synonym 6-Chloro-3-(1-methylethyl)-2,4(1H,3H)-pyrimidinedione, 6-Chloro-3-isopropylpyrimidine-2,4(1H,3H)-dione, Mavacamten Impurity
InChI formel InChI=1S/C7H9ClN2O2/c1-4(2)10-6(11)3-5(8)9-7(10)12/h3-4H,1-2H3,(H,9,12)
LEDER O=C1C=C(Cl)NC(=O)N1C(C)C
IUPAC-namn 6-klor-3-isopropylpyrimidin-2,4(1H,3H)-dion
Molekylvikt (g/mol) 188.61
Formel vikt 188.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.