missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-pyridinol, 97%, ACROS Organics™

Brand:  Acros Organics 379440050

Additional Details : CAS Number : 41288-96-4 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15205998

  • 2077.70 SEK / 5g


Estimated Shipment
06-11-2020
Add to basket


Specifications

Chemical Identifiers

41288-96-4
129.54
2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
6-chloropyridin-3-ol
C5H4ClNO
KVCOOWROABTXDJ-UHFFFAOYSA-N
819821
C1=CC(=NC=C1O)Cl

Specifications

6-Chloro-3-pyridinol
159.0°C to 161.0°C
41288-96-4
C5H4ClNO
KVCOOWROABTXDJ-UHFFFAOYSA-N
6-chloropyridin-3-ol
819821
97%
Authentic
5g
96% min. (HPLC)
2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
C1=CC(=NC=C1O)Cl
129.54
129.54
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Chloro-3-pyridinol, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents