Läs mer
6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone, Mikromol™
Produktkod. 17962506
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962506 25 mg
17902176 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17962506 Leverantör Mikromol Leverantörsnummer MM0071.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
CAS 23441-87-4
Analyt- eller komponentnamn 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
Frakt skick Room Temperature
Molekylformel C14 H8 Cl2 N2 O
Certifikat och efterlevnad ISO 17025
Synonym 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
InChI formel InChI=1S/C14H8Cl2N2O/c15-8-5-6-12-10(7-8)13(18-14(19)17-12)9-3-1-2-4-11(9)16/h1-7H,(H,17,18,19)
LEDER Clc1ccc2NC(=O)N=C(c3ccccc3Cl)c2c1
IUPAC-namn 6-chloro-4-(2-chlorophenyl)-1H-quinazolin-2-one
Molekylvikt (g/mol) 291.13
Kvantitet 25 mg
Formel vikt 290.0014 g/mol