Läs mer
6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid, TRC
Produktkod. 30005053
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005053 250 mg
30009123 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005053 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC364865250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54643-79-7
Molekylformel C15 H8 Cl2 N2 O2
Molekylvikt (g/mol) 319.14
Synonym 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid
IUPAC-namn 6-chloro-4-(2-chlorophenyl)quinazoline-2-carboxylic acid
LEDER OC(=O)c1nc(c2ccccc2Cl)c3cc(Cl)ccc3n1
Specifikationer
Kemiskt namn eller material 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C15H8Cl2N2O2/c16-8-5-6-12-10(7-8)13(19-14(18-12)15(20)21)9-3-1-2-4-11(9)17/h1-7H,(H,20,21)
Formel vikt 317.9963
Procent renhet >95%