Läs mer
6-Chloro-4-(2-chlorophenyl)-2-quinazolinemethanol, TRC
Produktkod. 30003943
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003943 10 mg
30010450 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30003943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36481510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 773871-49-1
Molekylformel C15 H10 Cl2 N2 O
Molekylvikt (g/mol) 305.16
Synonym 6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol
IUPAC-namn [6-chloro-4-(2-chlorophenyl)quinazolin-2-yl]methanol
LEDER OCc1nc(c2ccccc2Cl)c3cc(Cl)ccc3n1
Specifikationer
Kemiskt namn eller material 6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C15H10Cl2N2O/c16-9-5-6-13-11(7-9)15(19-14(8-20)18-13)10-3-1-2-4-12(10)17/h1-7,20H,8H2
Formel vikt 304.017
Procent renhet >95%