Läs mer
6-Chloro-4-hydroxy-2-methyl-2H-thieno[2,3-e]-1,2-thiazine-3-carboxylic Acid Methyl Ester 1,1-Dioxide, TRC
Produktkod. 30133869
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133869 25 g
30011948 1 g
30006862 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133869 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36663025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70415-50-8
Molekylformel C9H8ClNO5S2
Molekylvikt (g/mol) 309.75
Synonym Methyl 6-Chloro-4-hydroxy-2-methyl-2H-thieno[2,3-e]-1,2-thiazine-3-formate 1,1-Dioxide;
IUPAC-namn methyl 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-thieno[2,3-e]thiazine-3-carboxylate
LEDER COC(=O)C1=C(O)c2sc(Cl)cc2S(=O)(=O)N1C
Specifikationer
Kemiskt namn eller material 6-Chloro-4-hydroxy-2-methyl-2H-thieno[2,3-e]-1,2-thiazine-3-carboxylic Acid Methyl Ester 1,1-Dioxide
Kvantitet 25 g
InChI formel InChI=1S/C9H8ClNO5S2/c1-11-6(9(13)16-2)7(12)8-4(18(11,14)15)3-5(10)17-8/h3,12H,1-2H3
Formel vikt 308.9532