missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97+%, Thermo Scientific™

Catalog Number p-7025880
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18706-21-3
C10H5ClF3NO
247.601
MFCD00276583
MGEMMMJZAWYWNI-UHFFFAOYSA-N
6-chloro-4-hydroxy-2-trifluoromethyl quinoline, 6-chloro-2-trifluoromethyl quinolin-4-ol, 6-chloro-2-trifluoromethyl-1h-quinolin-4-one, 6-chloro-2-trifluoromethyl-quinolin-4-ol, 6-chloro-2-trifluoromethyl quinolin-4 1h-one, 6-chloro-2-trifluoromethylquinolin-4-ol, acmc-1boyc, 6-chloro-2-trifluoromethyl quinoline-4-ol, 2-trifluoromethyl-4-hydroxy-6-chloroquinoline
2736609
6-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)C(=O)C=C(N2)C(F)(F)F
This item is not returnable. View return policy
18706-21-3
C10H5ClF3NO
247.601
MFCD00276583
MGEMMMJZAWYWNI-UHFFFAOYSA-N
6-chloro-4-hydroxy-2-trifluoromethyl quinoline, 6-chloro-2-trifluoromethyl quinolin-4-ol, 6-chloro-2-trifluoromethyl-1h-quinolin-4-one, 6-chloro-2-trifluoromethyl-quinolin-4-ol, 6-chloro-2-trifluoromethyl quinolin-4 1h-one, 6-chloro-2-trifluoromethylquinolin-4-ol, acmc-1boyc, 6-chloro-2-trifluoromethyl quinoline-4-ol, 2-trifluoromethyl-4-hydroxy-6-chloroquinoline
2736609
6-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)C(=O)C=C(N2)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 278°C to 280°C
Quantity 1 g
Formula Weight 247.61
Percent Purity ≥97%
Chemical Name or Material 6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'