Läs mer
6-Chloro-6-deoxy-alpha-D-glucopyranose, TRC
Produktkod. 30359953
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359953 500 mg
30360700 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367760500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28528-86-1
Molekylformel C6H11ClO5
Molekylvikt (g/mol) 198.6
Synonym (2S,3R,4S,5S,6S)-6-(Chloromethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
IUPAC-namn (2S,3R,4S,5S,6S)-6-(chloromethyl)oxane-2,3,4,5-tetrol
LEDER O[C@H]1O[C@H](CCl)[C@@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material 6-Chloro-6-deoxy-alpha-D-glucopyranose
Kvantitet 500 mg
InChI formel InChI=1S/C6H11ClO5/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-6,8-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
Formel vikt 198.0295